Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCGGTCGTAGCCCCGGCCTCCTG[C/G]GGGTGGGATGGGGATTAAAAGATAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
46 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114280 MIM: 609151 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AIM1L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AIM1L - absent in melanoma 1-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039775.3 | Intron | NP_001034864.2 | ||||
XM_005245918.2 | Intron | XP_005245975.1 | ||||
XM_011541672.1 | Intron | XP_011539974.1 | ||||
XM_011541673.2 | Intron | XP_011539975.1 |
CD52 - CD52 molecule | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UBXN11 - UBX domain protein 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |