Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGACACCTCCTTTAAAGCCTCAC[A/G]GCGCCCCCATGAGGGAGGATCGTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
36 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600613 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MINOS1-NBL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MINOS1-NBL1 - MINOS1-NBL1 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204088.1 | Intron | NP_001191017.1 | ||||
NM_001204089.1 | Intron | NP_001191018.1 |
NBL1 - neuroblastoma 1, DAN family BMP antagonist | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204084.2 | Intron | NP_001191013.1 | ||||
NM_001204085.1 | Intron | NP_001191014.1 | ||||
NM_001204086.2 | Intron | NP_001191015.1 | ||||
NM_001278164.1 | Intron | NP_001265093.1 | ||||
NM_001278165.1 | Intron | NP_001265094.1 | ||||
NM_001278166.1 | Intron | NP_001265095.1 | ||||
NM_005380.7 | Intron | NP_005371.2 | ||||
NM_182744.3 | Intron | NP_877421.2 |