Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616519 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDE12 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PDE12 - phosphodiesterase 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322176.1 | 200 | Missense Mutation | CGG,TGG | R,W 23 | NP_001309105.1 | |
NM_001322177.1 | 200 | Missense Mutation | CGG,TGG | R,W 23 | NP_001309106.1 | |
NM_177966.6 | 200 | Missense Mutation | CGG,TGG | R,W 23 | NP_808881.3 |
Set Membership: |
HapMap |