Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCTGGGTGCTGAATCTCACTTCA[G/T]TGTTGACCCCCTTGGGGTTAGCATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616252 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TCAF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TCAF2 - TRPM8 channel associated factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130025.1 | 3090 | Intron | NP_001123497.1 | |||
NM_001130026.2 | 3090 | UTR 3 | NP_001123498.2 | |||
NM_173678.2 | 3090 | Intron | NP_775949.2 | |||
XM_006715927.3 | 3090 | UTR 3 | XP_006715990.1 | |||
XM_006715928.3 | 3090 | Intron | XP_006715991.1 | |||
XM_011516059.2 | 3090 | Intron | XP_011514361.1 |