Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCAAATAGTCAACTTTCACTGCCC[A/T]GTCATTTTAACCCACGTTTCAACAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 130590 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EEF1A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
EEF1A1 - eukaryotic translation elongation factor 1 alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001402.5 | 2120 | UTR 3 | NP_001393.1 | |||
XM_011535514.2 | 2120 | Intron | XP_011533816.1 |
Set Membership: |
Validated |