Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604134 MIM: 602930 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAMTS13 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ADAMTS13 - ADAM metallopeptidase with thrombospondin type 1 motif 13 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
REXO4 - REX4 homolog, 3'-5' exonuclease | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001279349.1 | Intron | NP_001266278.1 | ||||
NM_001279350.1 | Intron | NP_001266279.1 | ||||
NM_001279351.1 | Intron | NP_001266280.1 | ||||
NM_020385.3 | Intron | NP_065118.2 |
STKLD1 - serine/threonine kinase like domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |