Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604134 MIM: 602930 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAMTS13 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ADAMTS13 - ADAM metallopeptidase with thrombospondin type 1 motif 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_139025.4 | Intron | NP_620594.1 | ||||
NM_139026.4 | Intron | NP_620595.1 | ||||
NM_139027.4 | Intron | NP_620596.2 | ||||
XM_011518176.2 | Intron | XP_011516478.1 | ||||
XM_011518178.1 | Intron | XP_011516480.1 | ||||
XM_011518179.1 | Intron | XP_011516481.1 | ||||
XM_017014232.1 | Intron | XP_016869721.1 | ||||
XM_017014233.1 | Intron | XP_016869722.1 | ||||
XM_017014234.1 | Intron | XP_016869723.1 | ||||
XM_017014235.1 | Intron | XP_016869724.1 |
REXO4 - REX4 homolog, 3'-5' exonuclease | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001279349.1 | Intron | NP_001266278.1 | ||||
NM_001279350.1 | Intron | NP_001266279.1 | ||||
NM_001279351.1 | Intron | NP_001266280.1 | ||||
NM_020385.3 | Intron | NP_065118.2 |
Set Membership: |
HapMap Validated |