Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603531 MIM: 615783 MIM: 602186 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AP1S1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AP1S1 - adaptor related protein complex 1 sigma 1 subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NAT16 - N-acetyltransferase 16 (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198571.2 | 2195 | UTR 3 | NP_940973.2 | |||
XM_005250325.1 | 2195 | UTR 3 | XP_005250382.1 | |||
XM_011516168.1 | 2195 | UTR 3 | XP_011514470.1 |
VGF - VGF nerve growth factor inducible | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |