Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGGGAGAAACAGGACCAGACTGG[A/G]CGGCCACATTGGGCAGGCGCACGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
26 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605747 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LDLRAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LDLRAP1 - low density lipoprotein receptor adaptor protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015627.2 | 1081 | Intron | NP_056442.2 | |||
XM_006710559.3 | 1081 | Intron | XP_006710622.1 | |||
XM_006710560.3 | 1081 | Intron | XP_006710623.1 | |||
XM_006710561.3 | 1081 | Intron | XP_006710624.1 | |||
XM_011541209.2 | 1081 | Intron | XP_011539511.1 | |||
XM_011541210.2 | 1081 | Intron | XP_011539512.1 | |||
XM_011541211.2 | 1081 | Intron | XP_011539513.1 | |||
XM_011541212.2 | 1081 | Intron | XP_011539514.1 | |||
XM_017000994.1 | 1081 | UTR 5 | XP_016856483.1 | |||
XM_017000995.1 | 1081 | Intron | XP_016856484.1 |