Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612402 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALS2CL PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ALS2CL - ALS2 C-terminal like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190707.1 | 4297 | UTR 3 | NP_001177636.1 | |||
NM_147129.4 | 4297 | UTR 3 | NP_667340.2 | |||
XM_005265025.1 | 4297 | UTR 3 | XP_005265082.1 | |||
XM_006713091.2 | 4297 | Intron | XP_006713154.1 | |||
XM_006713093.3 | 4297 | Intron | XP_006713156.1 | |||
XM_006713094.3 | 4297 | Intron | XP_006713157.1 | |||
XM_011533572.2 | 4297 | Intron | XP_011531874.1 | |||
XM_017006120.1 | 4297 | Intron | XP_016861609.1 |