Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA1143 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KIAA1143 - KIAA1143 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KIF15 - kinesin family member 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020242.2 | Intron | NP_064627.1 | ||||
XM_006713264.3 | Intron | XP_006713327.1 | ||||
XM_011533964.2 | Intron | XP_011532266.1 | ||||
XM_017006884.1 | Intron | XP_016862373.1 | ||||
XM_017006885.1 | Intron | XP_016862374.1 | ||||
XM_017006886.1 | Intron | XP_016862375.1 | ||||
XM_017006887.1 | Intron | XP_016862376.1 | ||||
XM_017006888.1 | Intron | XP_016862377.1 | ||||
XM_017006889.1 | Intron | XP_016862378.1 | ||||
XM_017006890.1 | Intron | XP_016862379.1 |
Set Membership: |
HapMap |