Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGGTGTTTTAAGCTGTGAAAAAC[C/T]GAAGGCCAGCAGTATGGTCTGTATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615733 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJC30 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
DNAJC30 - DnaJ heat shock protein family (Hsp40) member C30 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WBSCR22 - Williams-Beuren syndrome chromosome region 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202560.2 | Intron | NP_001189489.1 | ||||
NM_017528.4 | Intron | NP_059998.2 | ||||
XM_006715847.1 | Intron | XP_006715910.1 | ||||
XM_011515778.1 | Intron | XP_011514080.1 | ||||
XM_011515779.1 | Intron | XP_011514081.1 |
Set Membership: |
HapMap Validated |