Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608074 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDCD6IP PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PDCD6IP - programmed cell death 6 interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001162429.2 | Intron | NP_001155901.1 | ||||
NM_001256192.1 | Intron | NP_001243121.1 | ||||
NM_013374.5 | Intron | NP_037506.2 | ||||
XM_011533252.1 | Intron | XP_011531554.1 | ||||
XM_011533253.1 | Intron | XP_011531555.1 | ||||
XM_017005488.1 | Intron | XP_016860977.1 |
Set Membership: |
HapMap |