Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGGGAAAGTGTGTATCCAGATTT[C/G]AAGACAGTGACTTTGCTTGGTTGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612202 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC102723313 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC102723313 - uncharacterized LOC102723313 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SOX7 - SRY-box 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031439.3 | 2515 | UTR 3 | NP_113627.1 |
Set Membership: |
HapMap |