Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATGCTGGGGTGAGGGTCCTGTGGT[A/G]TTGCTGGTGATGCTGACTGCCTTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610931 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZFAT PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ZFAT - zinc finger and AT-hook domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001029939.3 | 4263 | UTR 3 | NP_001025110.2 | |||
NM_001167583.2 | 4263 | UTR 3 | NP_001161055.1 | |||
NM_001174157.1 | 4263 | UTR 3 | NP_001167628.1 | |||
NM_001174158.1 | 4263 | UTR 3 | NP_001167629.1 | |||
NM_001289394.1 | 4263 | UTR 3 | NP_001276323.1 | |||
NM_020863.3 | 4263 | UTR 3 | NP_065914.2 | |||
XM_011517203.1 | 4263 | UTR 3 | XP_011515505.1 | |||
XM_011517204.2 | 4263 | UTR 3 | XP_011515506.1 | |||
XM_011517206.1 | 4263 | Intron | XP_011515508.1 | |||
XM_017013716.1 | 4263 | Intron | XP_016869205.1 |
Set Membership: |
HapMap Validated |