Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCAGTTGGCCAGGATGAGGCCATA[C/T]GCATGGTTCACACTTGGGGGGCCCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
49 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604468 MIM: 608042 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAP3K6 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MAP3K6 - mitogen-activated protein kinase kinase kinase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297609.1 | Intron | NP_001284538.1 | ||||
NM_004672.4 | Intron | NP_004663.3 | ||||
XM_017002771.1 | Intron | XP_016858260.1 | ||||
XM_017002772.1 | Intron | XP_016858261.1 |
SYTL1 - synaptotagmin like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |