Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
61 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607215 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR4689 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MIR4689 - microRNA 4689 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NPHP4 - nephronophthisis 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291593.1 | Intron | NP_001278522.1 | ||||
NM_001291594.1 | Intron | NP_001278523.1 | ||||
NM_015102.4 | Intron | NP_055917.1 | ||||
XM_006710563.3 | Intron | XP_006710626.1 | ||||
XM_011541213.1 | Intron | XP_011539515.1 | ||||
XM_011541214.1 | Intron | XP_011539516.1 | ||||
XM_011541215.1 | Intron | XP_011539517.1 | ||||
XM_011541216.2 | Intron | XP_011539518.1 | ||||
XM_011541217.2 | Intron | XP_011539519.1 | ||||
XM_011541218.2 | Intron | XP_011539520.1 | ||||
XM_017000996.1 | Intron | XP_016856485.1 | ||||
XM_017000997.1 | Intron | XP_016856486.1 | ||||
XM_017000998.1 | Intron | XP_016856487.1 | ||||
XM_017000999.1 | Intron | XP_016856488.1 | ||||
XM_017001000.1 | Intron | XP_016856489.1 | ||||
XM_017001001.1 | Intron | XP_016856490.1 | ||||
XM_017001002.1 | Intron | XP_016856491.1 | ||||
XM_017001003.1 | Intron | XP_016856492.1 |