Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607970 MIM: 614811 | ||||||||||||||||||||
Literature Links: |
GPR135 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GPR135 - G protein-coupled receptor 135 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
L3HYPDH - trans-L-3-hydroxyproline dehydratase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144581.1 | 2625 | UTR 3 | NP_653182.1 | |||
XM_005267298.3 | 2625 | UTR 3 | XP_005267355.1 | |||
XM_011536386.2 | 2625 | Intron | XP_011534688.1 | |||
XM_011536387.1 | 2625 | UTR 3 | XP_011534689.1 | |||
XM_011536388.1 | 2625 | UTR 3 | XP_011534690.1 |