Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607970 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR135 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | |||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
GPR135 - G protein-coupled receptor 135 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022571.5 | 7368 | Intron | NP_072093.2 | |||
XM_017021596.1 | 7368 | UTR 3 | XP_016877085.1 | |||
XM_017021597.1 | 7368 | UTR 3 | XP_016877086.1 | |||
XM_017021598.1 | 7368 | UTR 3 | XP_016877087.1 | |||
XM_017021599.1 | 7368 | UTR 3 | XP_016877088.1 |
Set Membership: |
HapMap Validated |