Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAGAAAGCATGAGAGGTTCTGACA[C/G]GGGGCACCCGTTTCATCTTGTCCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADCK5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADCK5 - aarF domain containing kinase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_174922.4 | Intron | NP_777582.4 | ||||
XM_006716527.2 | Intron | XP_006716590.1 | ||||
XM_011516907.2 | Intron | XP_011515209.1 | ||||
XM_011516909.1 | Intron | XP_011515211.1 | ||||
XM_011516910.1 | Intron | XP_011515212.1 | ||||
XM_017013174.1 | Intron | XP_016868663.1 |
LOC101928902 - uncharacterized LOC101928902 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |