Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605450 MIM: 612325 MIM: 606515 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GSTA4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | |||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
GSTA4 - glutathione S-transferase alpha 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ICK - intestinal cell (MAK-like) kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014920.3 | Intron | NP_055735.1 | ||||
NM_016513.4 | Intron | NP_057597.2 | ||||
XM_011514419.1 | Intron | XP_011512721.1 | ||||
XM_011514420.2 | Intron | XP_011512722.1 | ||||
XM_011514421.1 | Intron | XP_011512723.1 | ||||
XM_017010485.1 | Intron | XP_016865974.1 | ||||
XM_017010486.1 | Intron | XP_016865975.1 | ||||
XM_017010487.1 | Intron | XP_016865976.1 | ||||
XM_017010488.1 | Intron | XP_016865977.1 | ||||
XM_017010489.1 | Intron | XP_016865978.1 | ||||
XM_017010490.1 | Intron | XP_016865979.1 | ||||
XM_017010491.1 | Intron | XP_016865980.1 | ||||
XM_017010492.1 | Intron | XP_016865981.1 |
RN7SK - RNA, 7SK small nuclear | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |