Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTCCAGGCTCAGCCCTGCTCCTG[C/T]AGCTTTGCCGCTGAGTGTAGGAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616635 MIM: 603621 MIM: 615216 MIM: 609172 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYHR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CYHR1 - cysteine and histidine rich 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FOXH1 - forkhead box H1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003923.2 | 3155 | UTR 3 | NP_003914.1 |
KIFC2 - kinesin family member C2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145754.3 | 3155 | UTR 3 | NP_665697.1 | |||
XM_005272357.3 | 3155 | UTR 3 | XP_005272414.1 | |||
XM_011517362.2 | 3155 | Intron | XP_011515664.1 |
PPP1R16A - protein phosphatase 1 regulatory subunit 16A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |