Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGCCCTTGCCCCGCTCCGGCCGC[C/G]CTTCCCCGGGAGCCCTCAGGCTGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615093 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100288181 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC100288181 - uncharacterized LOC100288181 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LY6K - lymphocyte antigen 6 complex, locus K | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160354.1 | Intron | NP_001153826.1 | ||||
NM_001160355.1 | Intron | NP_001153827.1 | ||||
NM_017527.3 | Intron | NP_059997.3 | ||||
XM_005250989.4 | Intron | XP_005251046.1 | ||||
XM_005250990.4 | Intron | XP_005251047.1 |