Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTCGTTTGCATATCATTTGCATA[G/A]CATTTGCATGGACTTGTTTGTCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
37 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100288175 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC100288175 - uncharacterized LOC100288175 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105378948 - uncharacterized LOC105378948 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF223 - ring finger protein 223 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001205252.1 | Intron | NP_001192181.1 | ||||
XM_017001286.1 | Intron | XP_016856775.1 | ||||
XM_017001287.1 | Intron | XP_016856776.1 |