Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
33 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CFAP74 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CFAP74 - cilia and flagella associated protein 74 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304360.1 | 2790 | Missense Mutation | AGG,GGG | R,G 1069 | NP_001291289.1 | |
XM_011542333.2 | 2790 | Missense Mutation | AGG,GGG | R,G 1092 | XP_011540635.2 | |
XM_011542335.1 | 2790 | Missense Mutation | AGG,GGG | R,G 900 | XP_011540637.1 | |
XM_011542336.2 | 2790 | Missense Mutation | AGG,GGG | R,G 900 | XP_011540638.1 | |
XM_017002641.1 | 2790 | Missense Mutation | AGG,GGG | R,G 1111 | XP_016858130.1 | |
XM_017002642.1 | 2790 | Missense Mutation | AGG,GGG | R,G 1111 | XP_016858131.1 | |
XM_017002643.1 | 2790 | Missense Mutation | AGG,GGG | R,G 1111 | XP_016858132.1 | |
XM_017002644.1 | 2790 | Missense Mutation | AGG,GGG | R,G 255 | XP_016858133.1 |