Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 176891 | ||||||||||||||||||||
Literature Links: |
PTPRZ1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PTPRZ1 - protein tyrosine phosphatase, receptor type Z1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206838.1 | 324 | UTR 5 | NP_001193767.1 | |||
NM_001206839.1 | 324 | UTR 5 | NP_001193768.1 | |||
NM_002851.2 | 324 | UTR 5 | NP_002842.2 | |||
XM_005250519.1 | 324 | UTR 5 | XP_005250576.1 | |||
XM_017012477.1 | 324 | UTR 5 | XP_016867966.1 |