Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGAGTGGGGAGAACAGGCTTTGCC[G/A]GACCCCAGGACCCACGTAAGACACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
40 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
MORN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MORN1 - MORN repeat containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301060.1 | Intron | NP_001287989.1 | ||||
NM_024848.2 | Intron | NP_079124.1 | ||||
XM_005244798.2 | Intron | XP_005244855.1 | ||||
XM_011542169.1 | Intron | XP_011540471.1 | ||||
XM_011542170.2 | Intron | XP_011540472.1 | ||||
XM_011542172.2 | Intron | XP_011540474.1 | ||||
XM_011542173.2 | Intron | XP_011540475.1 | ||||
XM_011542174.2 | Intron | XP_011540476.1 | ||||
XM_011542176.1 | Intron | XP_011540478.1 | ||||
XM_011542177.1 | Intron | XP_011540479.1 | ||||
XM_017002365.1 | Intron | XP_016857854.1 |