Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGCCATCTTCCCTGTATACAGAC[A/G]AGGCAGTTGAGGTCCAGGGAAAGTC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606055 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PCTP PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
PCTP - phosphatidylcholine transfer protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102402.2 | Intron | NP_001095872.1 | ||||
NM_021213.3 | Intron | NP_067036.2 | ||||
XM_005257549.1 | Intron | XP_005257606.1 | ||||
XM_005257550.3 | Intron | XP_005257607.1 | ||||
XM_017024913.1 | Intron | XP_016880402.1 |