Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300065 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CENPI PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CENPI - centromere protein I | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318521.1 | Intron | NP_001305450.1 | ||||
NM_001318523.1 | Intron | NP_001305452.1 | ||||
NM_006733.3 | Intron | NP_006724.2 | ||||
XM_005262111.2 | Intron | XP_005262168.1 | ||||
XM_011530894.2 | Intron | XP_011529196.1 | ||||
XM_011530895.2 | Intron | XP_011529197.1 | ||||
XM_011530897.2 | Intron | XP_011529199.1 | ||||
XM_011530899.2 | Intron | XP_011529201.1 | ||||
XM_017029383.1 | Intron | XP_016884872.1 |
TMEM35A - transmembrane protein 35A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |