Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300191 | ||||||||||||||||||||
Literature Links: |
TSPAN6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TSPAN6 - tetraspanin 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278740.1 | 1932 | UTR 3 | NP_001265669.1 | |||
NM_001278741.1 | 1932 | UTR 3 | NP_001265670.1 | |||
NM_001278742.1 | 1932 | UTR 3 | NP_001265671.1 | |||
NM_001278743.1 | 1932 | UTR 3 | NP_001265672.1 | |||
NM_003270.3 | 1932 | UTR 3 | NP_003261.1 | |||
XM_011531018.1 | 1932 | UTR 3 | XP_011529320.1 |