Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAATTCGACTTTTCTGAATCTAAT[C/A]GGAAAATAAGTGCCCCGTTCCCCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608463 MIM: 601999 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AATF PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
AATF - apoptosis antagonizing transcription factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LHX1 - LIM homeobox 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005568.4 | Intron | NP_005559.2 |
LOC102723471 - uncharacterized LOC102723471 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |