Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
66 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606077 MIM: 603878 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RBM15 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RBM15 - RNA binding motif protein 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201545.1 | Intron | NP_001188474.1 | ||||
NM_022768.4 | Intron | NP_073605.4 |
SLC16A4 - solute carrier family 16 member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201546.1 | Intron | NP_001188475.1 | ||||
NM_001201547.1 | Intron | NP_001188476.1 | ||||
NM_001201548.1 | Intron | NP_001188477.1 | ||||
NM_001201549.1 | Intron | NP_001188478.1 | ||||
NM_001319220.1 | Intron | NP_001306149.1 | ||||
NM_004696.2 | Intron | NP_004687.1 | ||||
XM_005271317.4 | Intron | XP_005271374.1 | ||||
XM_006711033.3 | Intron | XP_006711096.1 |
Set Membership: |
HapMap |