Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTACAGAAATAGACAATAGAATC[C/T]TAAAATTCACATGGGATGACAAAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 601968 | ||||||||||||||||||||
Literature Links: |
WNT2B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
WNT2B - Wnt family member 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291880.1 | Intron | NP_001278809.1 | ||||
NM_004185.4 | Intron | NP_004176.2 | ||||
NM_024494.2 | Intron | NP_078613.1 |