Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAATGCCTTTCCCAATAAAACTCGT[G/T]AACAAAATAGTGGTAACATATAAAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
48 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615464 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDX59 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
DDX59 - DEAD-box helicase 59 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031725.5 | Intron | NP_001026895.2 | ||||
NM_001320181.1 | Intron | NP_001307110.1 | ||||
NM_001320182.1 | Intron | NP_001307111.1 | ||||
XM_005245519.1 | Intron | XP_005245576.1 | ||||
XM_005245521.2 | Intron | XP_005245578.1 | ||||
XM_006711562.1 | Intron | XP_006711625.1 | ||||
XM_011510035.2 | Intron | XP_011508337.1 | ||||
XM_017002431.1 | Intron | XP_016857920.1 | ||||
XM_017002432.1 | Intron | XP_016857921.1 |