Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCAAGGGACTTGTTTGGGGCCCTC[G/T]CTTATCTGTGCCTCAGTTTCCCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 155120 MIM: 611661 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAM11 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ADAM11 - ADAM metallopeptidase domain 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318933.1 | Intron | NP_001305862.1 | ||||
NM_002390.5 | Intron | NP_002381.2 | ||||
XM_005257373.3 | Intron | XP_005257430.1 | ||||
XM_017024647.1 | Intron | XP_016880136.1 |
DBF4B - DBF4 zinc finger B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |