Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601029 MIM: 607794 MIM: 611768 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MEST PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MEST - mesoderm specific transcript | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253900.1 | Intron | NP_001240829.1 | ||||
NM_001253901.1 | Intron | NP_001240830.1 | ||||
NM_001253902.1 | Intron | NP_001240831.1 | ||||
NM_002402.3 | Intron | NP_002393.2 | ||||
NM_177524.2 | Intron | NP_803490.1 | ||||
NM_177525.2 | Intron | NP_803491.1 | ||||
XM_011516222.1 | Intron | XP_011514524.1 | ||||
XM_017012218.1 | Intron | XP_016867707.1 |
MESTIT1 - MEST intronic transcript 1, antisense RNA | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR335 - microRNA 335 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |