Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAGTGTTACACTTGAAAAAATTCC[C/T]TAAAATATAGTATTACTTAGATCAA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 605073 | |||||||||||||||||||||||
Literature Links: |
PPM1E PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
PPM1E - protein phosphatase, Mg2+/Mn2+ dependent 1E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014906.4 | Intron | NP_055721.3 | ||||
XM_005257169.4 | Intron | XP_005257226.1 | ||||
XM_011524534.2 | Intron | XP_011522836.1 | ||||
XM_017024359.1 | Intron | XP_016879848.1 |
Set Membership: |
HapMap |