Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605752 MIM: 607827 MIM: 607828 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HID1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HID1 - HID1 domain containing | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OTOP2 - otopetrin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OTOP3 - otopetrin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001272005.1 | 257 | Missense Mutation | CAG,CGG | Q,R 46 | NP_001258934.1 | |
NM_178233.2 | 257 | Missense Mutation | CAG,CGG | Q,R 64 | NP_839947.1 | |
XM_011524744.2 | 257 | Missense Mutation | CAG,CGG | Q,R 35 | XP_011523046.1 |
Set Membership: |
HapMap |