Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGGAGAAAACTTGTAACCATCCC[C/T]GACATAGAACTTGTTCTGGAACTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605239 MIM: 611700 | ||||||||||||||||||||
Literature Links: |
ATP6V0A4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATP6V0A4 - ATPase H+ transporting V0 subunit a4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020632.2 | 2696 | Missense Mutation | AGG,GGG | R,G 820 | NP_065683.2 | |
NM_130840.2 | 2696 | Missense Mutation | AGG,GGG | R,G 820 | NP_570855.2 | |
NM_130841.2 | 2696 | Missense Mutation | AGG,GGG | R,G 820 | NP_570856.2 | |
XM_005250393.1 | 2696 | Missense Mutation | AGG,GGG | R,G 820 | XP_005250450.1 | |
XM_005250394.3 | 2696 | Missense Mutation | AGG,GGG | R,G 820 | XP_005250451.1 |
SVOPL - SVOP like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |