Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAATATAATTATAAAACCAAGTCA[C/G]AAATACAAAAAAAGGGACAACTATA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602208 MIM: 610273 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNJ10 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
KCNJ10 - potassium voltage-gated channel subfamily J member 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PIGM - phosphatidylinositol glycan anchor biosynthesis class M | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145167.2 | 3785 | UTR 3 | NP_660150.1 |
Set Membership: |
HapMap |