Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAGTTACTATCTATAGAGAAATAC[A/G]AAAAGACATTTTATCCATGAAATGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615169 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AMZ2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
AMZ2 - archaelysin family metallopeptidase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033569.1 | Intron | NP_001028741.1 | ||||
NM_001033570.1 | Intron | NP_001028742.1 | ||||
NM_001033571.1 | Intron | NP_001028743.1 | ||||
NM_001033572.1 | Intron | NP_001028744.1 | ||||
NM_001033574.1 | Intron | NP_001028746.1 | ||||
NM_001289054.1 | Intron | NP_001275983.1 | ||||
NM_001289056.1 | Intron | NP_001275985.1 | ||||
NM_016627.4 | Intron | NP_057711.3 | ||||
XM_005257432.4 | Intron | XP_005257489.1 | ||||
XM_005257433.3 | Intron | XP_005257490.1 | ||||
XM_005257435.4 | Intron | XP_005257492.1 | ||||
XM_005257436.3 | Intron | XP_005257493.1 | ||||
XM_006721940.2 | Intron | XP_006722003.1 | ||||
XM_017024723.1 | Intron | XP_016880212.1 | ||||
XM_017024724.1 | Intron | XP_016880213.1 | ||||
XM_017024725.1 | Intron | XP_016880214.1 | ||||
XM_017024726.1 | Intron | XP_016880215.1 | ||||
XM_017024727.1 | Intron | XP_016880216.1 | ||||
XM_017024728.1 | Intron | XP_016880217.1 |