Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
68 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138290 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLUL PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GLUL - glutamate-ammonia ligase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LINC00272 - long intergenic non-protein coding RNA 272 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TEDDM1 - transmembrane epididymal protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_172000.3 | 674 | Silent Mutation | CCC,CCT | P,P 181 | NP_741997.3 |