Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCGCTCGCGGGCCCCACGTTCAGG[C/T]AGCGCAGGCACCAAAAGTTTCCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609391 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC01124 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LINC01124 - long intergenic non-protein coding RNA 1124 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC101926913 - uncharacterized LOC101926913 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SP5 - Sp5 transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003845.2 | 1346 | Intron | NP_001003845.1 | |||
XM_005246542.4 | 1346 | Intron | XP_005246599.1 | |||
XM_011511159.2 | 1346 | Missense Mutation | GCA,GTA | A,V 62 | XP_011509461.1 |