Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGCTGTTCTAGGTTCAGTGTTTGG[C/T]TTTTAACCTTACCTTTTCTTTTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607185 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEC24C PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SEC24C - SEC24 homolog C, COPII coat complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004922.3 | Intron | NP_004913.2 | ||||
NM_198597.2 | Intron | NP_940999.1 | ||||
XM_011540380.2 | Intron | XP_011538682.1 | ||||
XM_011540381.2 | Intron | XP_011538683.1 | ||||
XM_017016967.1 | Intron | XP_016872456.1 | ||||
XM_017016968.1 | Intron | XP_016872457.1 |
Set Membership: |
HapMap |