Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601572 MIM: 614760 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAPZB PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CAPZB - capping actin protein of muscle Z-line beta subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206540.2 | 1495 | UTR 3 | NP_001193469.1 | |||
NM_001206541.2 | 1495 | UTR 3 | NP_001193470.1 | |||
NM_001282162.1 | 1495 | UTR 3 | NP_001269091.1 | |||
NM_001313932.1 | 1495 | Intron | NP_001300861.1 | |||
NM_004930.4 | 1495 | UTR 3 | NP_004921.1 | |||
XM_006710938.3 | 1495 | Intron | XP_006711001.1 | |||
XM_011542228.2 | 1495 | Intron | XP_011540530.1 | |||
XM_011542229.2 | 1495 | Intron | XP_011540531.1 | |||
XM_011542230.2 | 1495 | Intron | XP_011540532.1 | |||
XM_017002428.1 | 1495 | Intron | XP_016857917.1 | |||
XM_017002429.1 | 1495 | Intron | XP_016857918.1 | |||
XM_017002430.1 | 1495 | Intron | XP_016857919.1 |
PQLC2 - PQ loop repeat containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |