Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
23 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602234 MIM: 609444 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CASP9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CASP9 - caspase 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001229.4 | 1272 | Intron | NP_001220.2 | |||
NM_001278054.1 | 1272 | Intron | NP_001264983.1 | |||
NM_032996.3 | 1272 | Intron | NP_127463.2 | |||
XM_005246014.2 | 1272 | Intron | XP_005246071.1 | |||
XM_011542273.2 | 1272 | Missense Mutation | XP_011540575.1 |
CELA2B - chymotrypsin like elastase family member 2B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |