Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTGTTCTATTTTGCTCTTTGAAT[A/G]GATCTTTTTACCTAGATGTGATTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604993 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRPF18 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PRPF18 - pre-mRNA processing factor 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003675.3 | Intron | NP_003666.1 | ||||
XM_011519754.2 | Intron | XP_011518056.1 | ||||
XM_017016860.1 | Intron | XP_016872349.1 |
Set Membership: |
HapMap |