Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610103 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
S100Z PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
S100Z - S100 calcium binding protein Z | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_130772.3 | Intron | NP_570128.2 | ||||
XM_011543239.2 | Intron | XP_011541541.1 | ||||
XM_011543240.2 | Intron | XP_011541542.1 | ||||
XM_011543241.2 | Intron | XP_011541543.1 | ||||
XM_011543244.2 | Intron | XP_011541546.1 | ||||
XM_011543245.2 | Intron | XP_011541547.1 | ||||
XM_017009171.1 | Intron | XP_016864660.1 | ||||
XM_017009172.1 | Intron | XP_016864661.1 | ||||
XM_017009173.1 | Intron | XP_016864662.1 |
Set Membership: |
HapMap |