Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602968 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCAS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
BCAS1 - breast carcinoma amplified sequence 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316361.2 | 2649 | UTR 3 | NP_001303290.1 | |||
NM_001323347.1 | 2649 | UTR 3 | NP_001310276.1 | |||
NM_003657.3 | 2649 | UTR 3 | NP_003648.2 | |||
XM_005260587.2 | 2649 | UTR 3 | XP_005260644.1 | |||
XM_005260589.2 | 2649 | UTR 3 | XP_005260646.1 | |||
XM_005260590.2 | 2649 | UTR 3 | XP_005260647.1 | |||
XM_005260591.2 | 2649 | UTR 3 | XP_005260648.1 | |||
XM_005260593.2 | 2649 | UTR 3 | XP_005260650.1 | |||
XM_005260594.2 | 2649 | UTR 3 | XP_005260651.1 | |||
XM_005260595.2 | 2649 | UTR 3 | XP_005260652.1 | |||
XM_011529090.2 | 2649 | UTR 3 | XP_011527392.1 | |||
XM_011529091.2 | 2649 | UTR 3 | XP_011527393.1 | |||
XM_017028110.1 | 2649 | UTR 3 | XP_016883599.1 | |||
XM_017028111.1 | 2649 | UTR 3 | XP_016883600.1 | |||
XM_017028112.1 | 2649 | UTR 3 | XP_016883601.1 |