Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600509 MIM: 600937 MIM: 613714 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCC8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABCC8 - ATP binding cassette subfamily C member 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KCNJ11 - potassium voltage-gated channel subfamily J member 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000525.3 | 1462 | UTR 3 | NP_000516.3 | |||
NM_001166290.1 | 1462 | UTR 3 | NP_001159762.1 | |||
XM_006718226.3 | 1462 | UTR 3 | XP_006718289.1 | |||
XM_017017680.1 | 1462 | UTR 3 | XP_016873169.1 |
NCR3LG1 - natural killer cell cytotoxicity receptor 3 ligand 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |